Learn More
H60 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1155
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13456774
  • £360.15 / 100µg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
H60 Polyclonal antibody AF1155 Specifically detects H60 in Mouse samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications
Specifications
H60
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction
Q3TDZ7
H60a
Mouse myeloma cell line NS0-derived recombinant mouse H60 Asp30-Gln212 Accession # Q3TDZ7
Antigen Affinity-purified
RUO
Primary
15101
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse H60 in direct ELISAs and Western blots.