missing translation for 'onlineSavingsMsg'
Learn More

Glypican 5 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2689

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447724

  • £344.84 / 100µg

Estimated Shipment
15-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Glypican 5 Polyclonal antibody AF2689 Specifically detects Glypican 5 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Glypican 5
LYOPH
Western Blot 0.1 ug/mL
Q8CAL5
bA93M14.1, glypican 5, glypican proteoglycan 5, glypican-5, GPC5
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Glypican 5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant human (rh) Glypican 5 is observed and less than 1% cross-reactivity with recombinant mouse Glypican 2, rhGlypican 3, and rhGlypican 6 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
GPC5
Mouse myeloma cell line NS0-derived recombinant mouse Glypican 5 Gly26-Gly549 Accession # Q8CAL5
Immunogen affinity purified
RUO
Primary
2262
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only