missing translation for 'onlineSavingsMsg'
Learn More

GFR alpha-4/GDNF R alpha-4 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1677

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467084

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 26-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

GFR alpha-4/GDNF R alpha-4 Polyclonal antibody AF1677 Specifically detects GFR alpha-4/GDNF R alpha-4 in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

GFR alpha-4/GDNF R alpha-4/Persephin receptor
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Affinity Purified
GFRA4
Mouse myeloma cell line NS0-derived recombinant mouse GFR alpha-4/GDNF R alpha-4 Asn24-Thr237 Accession # Q99JT2
Immunogen affinity purified
RUO
Primary
64096
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF R alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR alpha4, GFR receptor alpha 4, GFRa-4, GFRA4, GFR-alpha-4, Persephin receptor
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse GFR alpha-4/GDNF R alpha-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human GFR alpha-4 is observed.
Mouse
Videos
Documents

For Research Use Only