missing translation for 'onlineSavingsMsg'
Learn More

GDF-1 Propeptide Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3069

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487954

  • £357.73 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

GDF-1 Propeptide Polyclonal antibody AF3069 Specifically detects GDF-1 Propeptide in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

GDF-1
LYOPH
Western Blot 0.1 ug/mL
P20863
DORV, DTGA3, embryonic growth/differentiation factor 1, GDF1, GDF-1, growth differentiation factor 1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse GDF-1 Propeptide in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with mature recombinant mouse (rm) GDF-1 and rmGDF-3 Propeptide is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
GDF1
Mouse myeloma cell line NS0-derived recombinant mouse GDF-1 Propeptide, Pro25-Leu235, Accession # P20863
Immunogen affinity purified
RUO
Primary
2657
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only