missing translation for 'onlineSavingsMsg'
Learn More

FGF-6 Sheep anti-Mouse, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF5750

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13439414

  • £304.93 / 100µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FGF-6 Polyclonal antibody AF5750 Specifically detects FGF-6 in Mouse samples. It is validated for Immunohistochemistry
Specifications

Specifications

FGF-6
LYOPH
Immunohistochemistry 5-15 ug/mL
NP_034334
FGF6, FGF-6, fibroblast growth factor 6, HBGF-6, Heparin secretory-transforming protein 2, Heparin-binding growth factor 6, HST2, HST-2, HSTF2, HSTF-2
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse FGF-6 in direct ELISAs. In direct ELISAs, approximately 15% cross-reactivity with recombinant human FGF-6 and less than 1 % cross-reactivity with recombinant mouse FGF-7 is observed.
Mouse
Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FGF6
E. coli-derived recombinant mouse FGF-6 Pro39-Ile208 Accession # NP_034334
Antigen Affinity-purified
RUO
Primary
2251
Sterile PBS to a final concentration of 0.2 mg/mL.
Videos

For Research Use Only