missing translation for 'onlineSavingsMsg'
Learn More

FGF-4 Sheep anti-Mouse, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF5846

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449464

  • £339.57 / 100µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FGF-4 Polyclonal antibody AF5846 Specifically detects FGF-4 in Mouse samples. It is validated for Immunohistochemistry
Specifications

Specifications

FGF-4
LYOPH
Immunohistochemistry 5-15 ug/mL
P11403
FGF4, fibroblast growth factor 4, HBGF-4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse FGF-4 in direct ELISAs. In direct ELISAs, approximately 60% cross-reactivity with recombinant human FGF-4 is observed.
Mouse
Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FGF4
E. coli-derived recombinant mouse FGF-4 Ala30-Leu202 Accession # P11403
Immunogen affinity purified
RUO
Primary
2249
Sterile PBS to a final concentration of 0.2 mg/mL.
Videos
Documents

For Research Use Only