missing translation for 'onlineSavingsMsg'
Learn More

Fc gamma RIII (CD16) Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1960

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457254

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Fc gamma RIII (CD16) Polyclonal antibody AF1960 Specifically detects Fc gamma RIII (CD16) in Mouse samples. It is validated for Blocking (Receptor-ligand), Immunocytochemistry, Western Blotting
Specifications

Specifications

Fc gamma RIII (CD16)
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction, Immunocytochemistry 5-15 ug/mL
P08508
CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects Fc gamma RIIIA/B (CD16) in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) Fc gamma RIIIB and rhCD32 is observed.
Mouse
Western Blot, Blockade of Receptor-ligand Interaction, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Fcgr3
Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIII Ala31-Thr215 Accession # P08508
Immunogen affinity purified
RUO
Primary
2214
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only