missing translation for 'onlineSavingsMsg'
Learn More

Fc gamma RI/CD64 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2074

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487324

  • £377.53 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Fc gamma RI/CD64 Polyclonal antibody AF2074 Specifically detects Fc gamma RI/CD64 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Fc gamma RI/CD64
LYOPH
Western Blot 0.5 ug/mL
P26151
CD64, CD64 antigen, CD64b, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FCGR1B, FcgRI, FCRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse Fc gamma RI/CD64 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human Fc gamma RI is observed and less than 5% cross-reactivity with recombinant mouse (rm) Fc gamma RIIB and rmFc gamma RIII is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Fcgr1
NS0-derived recombinant mouse Fc gamma RI/CD64, Glu25-Pro297, Accession # P26151
Antigen Affinity-purified
RUO
Primary
2209
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only