missing translation for 'onlineSavingsMsg'
Learn More

FAM3D Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3027

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13427934

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FAM3D Polyclonal antibody AF3027 Specifically detects FAM3D in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

FAM3D
LYOPH
Western Blot 0.1 ug/mL
P97805
cytokine-like protein EF-7, EF7, family with sequence similarity 3, member D, OIT1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse FAM3D in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) FAM3D is observed and less than 1% cross reactivity with rhFAM3A, rhFAM3B and rhFAM3C is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FAM3D
Mouse myeloma cell line NS0-derived recombinant mouse FAM3D Trp39-Met224 Accession # P97805
Immunogen affinity purified
RUO
Primary
131177
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only