missing translation for 'onlineSavingsMsg'
Learn More

FAM3B Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2867

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487834

  • £377.53 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

FAM3B Polyclonal antibody AF2867 Specifically detects FAM3B in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

FAM3B
LYOPH
Western Blot 0.1 ug/mL
Q9D309
2-21, C21orf11, C21orf76PANDER, chromosome 21 open reading frame 11, Cytokine-like protein 2-21, D21M16SJHU19e, family with sequence similarity 3, member B, ORF9, pancreatic derived factor, Pancreatic-derived factor, PANDER, PRED44
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse FAM3B in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human FAM3B is observed, approximately 5% cross-reactivity with recombinant mouse (rm) FAM3C is observed, and less than 1% cross-reactivity with rmFAM3D is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FAM3B
Mouse myeloma cell line NS0-derived recombinant mouse FAM3B, Glu30-Arg235, Accession # Q9D309
Immunogen affinity purified
RUO
Primary
54097
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only