missing translation for 'onlineSavingsMsg'
Learn More

Exostosin-like 2/EXTL2 Lumenal Domain Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2536

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437624

  • £332.22 / 100µg

Estimated Shipment date
from Supplier 23-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Exostosin-like 2/EXTL2 Lumenal Domain Polyclonal antibody AF2536 Specifically detects Exostosin-like 2/EXTL2 Lumenal Domain in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Exostosin-like 2/EXTL2
LYOPH
Western Blot 0.1 ug/mL
Q9ES89
Alpha-1,4-N-acetylhexosaminyltransferase EXTL2, alpha-1,4-N-acteylhexosaminyltransferase, Alpha-GalNAcT EXTL2, EC 2.4.1.223, exostoses (multiple)-like 2, Exostosin like 2, exostosin-like 2, EXTL2, EXTR2, EXT-related protein 2, Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, processed exostosin-like 2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Exostosin-like 2/EXTL2 Lumenal Domain in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse EXTL1 and recombinant human EXTL3 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
EXTL2
Mouse myeloma cell line NS0-derived recombinant mouse Exostosin-like 2/EXTL2 Lumenal Domain Asn43-Met330 Accession # Q9ES89
Antigen Affinity-purified
RUO
Primary
2135
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only