missing translation for 'onlineSavingsMsg'
Learn More

Ephrin-A1 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF702

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13469964

  • £454.23 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ephrin-A1 Polyclonal antibody AF702 Specifically detects Ephrin-A1 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Ephrin-A1
LYOPH
Western Blot 0.1 ug/mL
P52793
B61, ECKLG, EFL1, EFL-1, EFNA1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EphrinA1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Ephrin-A1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) Ephrin-A1 is observed and less than 1% cross-reactivity with recombinant mouse (rm) Ephrin-B1, rmEphrin-A5, rmEphrin-A4, rmEphrin-A2, and rmEphrin-B1 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
EFNA1
Mouse myeloma cell line NS0-derived recombinant mouse Ephrin-A1 Asp19-Ser182 Accession # P52793
Antigen Affinity-purified
RUO
Primary
1942
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only