missing translation for 'onlineSavingsMsg'
Learn More

EphA8 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF454

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458784

  • £377.53 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

EphA8 Polyclonal antibody AF454 Specifically detects EphA8 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

EphA8
LYOPH
Western Blot 0.1 ug/mL
O09127
EEK, EK3, EPH- and ELK-related kinase, EPH- and ELK-related tyrosine kinase, EPH receptor A8, EPH-like kinase 3, ephrin type-A receptor 8, HEK3, hydroxyaryl-protein kinase, KIAA1459, protein-tyrosine kinase, tyrosine-protein kinase receptor EEK, tyrosylprotein kinase
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects recombinant mouse EphA8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant rat EphA5, recombinant mouse (rm) EphA6, rmEphA7, rmEphA2, recombinant human EphA1, and rmEphA3 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
EPHA8
Mouse myeloma cell line NS0-derived recombinant mouse EphA8, Gly27 - Arg540, Accession # O09127
Antigen Affinity-purified
RUO
Primary
2046
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only