missing translation for 'onlineSavingsMsg'
Learn More

EphA7 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF608

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13439584

  • £314.17 / 100µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

EphA7 Polyclonal antibody AF608 Specifically detects EphA7 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

EphA7
LYOPH
Western Blot 0.1 ug/mL
Q61772
Cek11, Ebk, EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, Mdk1, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse EphA7 in direct ELISAs and Western blots. In direct ELISAs, approximately 2% cross-reactivity with recombinant mouse (rm) EphA3 is observed and less than 1% cross-reactivity with rmEphA2, rmEphA4, rmEphA6, and rmEphA8 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
EPHA7
Mouse myeloma cell line NS0-derived recombinant mouse EphA7 isoform 1 Ala30-Pro554 (Gly540-Glu544 del) Accession # Q61772
Immunogen affinity purified
RUO
Primary
2045
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only