missing translation for 'onlineSavingsMsg'
Learn More

Desert Hedgehog/Dhh N-Terminus Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF733

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440085

  • £298.33 / 100µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Desert Hedgehog/Dhh N-Terminus Polyclonal antibody AF733 Specifically detects Desert Hedgehog/Dhh N-Terminus in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Desert Hedgehog/Dhh
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Q61488
desert hedgehog, desert hedgehog (Drosophila) homolog, desert hedgehog homolog, desert hedgehog protein, DHH, HHG-3MGC35145
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Desert Hedgehog/Dhh N-Terminus in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse (rm) Shh N-terminal peptide (aa 25-198) is observed. Additionally, in direct ELISAs, no cross-reactivity with rmDhh C-terminal peptide (aa 199-396) and rmIhh C-terminal peptide (aa 241-449) is observed.
Mouse
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
DHH
E. coli-derived recombinant mouse Desert Hedgehog/Dhh aa 23-198 Accession # Q61488
Antigen Affinity-purified
RUO
Primary
50846
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only