Learn More
DcTRAIL R1/TNFRSF23 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2378
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487514
  • £382.00 / 100µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
DcTRAIL R1/TNFRSF23 Polyclonal antibody AF2378 specifically detects DcTRAIL R1/TNFRSF23 in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
DcTRAIL R1/TNFRSF23
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse DcTRAIL R1/TNFRSF23, Met30-Ser156, Accession No. Q9ER63
100μg
Primary
79201
Unconjugated
Q9ER63
Goat
IgG
RUO
Polyclonal
Murine
Videos