Learn More
DCAR/CLEC4B Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2757
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487764
  • £377.53 / 100µg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
DCAR/CLEC4B Polyclonal antibody AF2757 Specifically detects DCAR/CLEC4B in Mouse samples. It is validated for Flow Cytometry, Western Blotting
Specifications
Specifications
DCAR/CLEC4B
Unconjugated
Q7TS58
Goat
IgG
RUO
Polyclonal
Murine
CyTOF, Flow Cytometry, Western Blotting
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse DCAR/CLEC4B, Gln42-Leu209, Accession # Q7TS58
100μg
Primary
69810