Learn More
DAN Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF755
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13430145
  • £362.60 / 100µg
Estimated Shipment
14-09-2020
Description
Description
DAN Polyclonal antibody AF755 Specifically detects DAN in Mouse samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications
Specifications
DAN/NBL1
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
Q499K5
DAN domain family member 1, DAND1, MGC8972, NBL1, neuroblastoma suppressor of tumorigenicity 1, neuroblastoma, suppression of tumorigenicity 1, Protein N03, suppression of tumorigenicity 1, Zinc finger protein DAN
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse DAN in direct ELISAs and Western blots.
Mouse
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NBL1
Mouse myeloma cell line NS0-derived recombinant mouse DAN Ala17-Asp178 Accession # Q499K5
Immunogen affinity purified
RUO
Primary
4681
Reconstitute at 0.2 mg/mL in sterile PBS.