missing translation for 'onlineSavingsMsg'
Learn More

CXCL15/Lungkine Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF442

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13428724

  • £342.51 / 100µg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CXCL15/Lungkine Polyclonal antibody AF442 Specifically detects CXCL15/Lungkine in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

CXCL15/Lungkine
LYOPH
Western Blot 0.1 ug/mL
Q9WVL7
lungkine, Scyb15, weche
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse CXCL15/Lungkine in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with rm6Ckine, rmC10, rrCINC-1, rmBLC, rmCRG-2, rmEotaxin, rmFractalkine, rmGCP-2, rmJE, rmKC, rmLymphotactin, rmMARC, rmMDC, rmMIG, rmMIP-1 alpha , rmMIP-1 beta , rmRANTES, rmSDF-1 alpha , rmTCA-3 and rmTeck is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CXCL15
E. coli-derived recombinant mouse CXCL15/Lungkine (R&D Systems, Catalog # 442-LK) Gln26-Ala167 Accession # Q9WVL7
Immunogen affinity purified
RUO
Primary
20309
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only