missing translation for 'onlineSavingsMsg'
Learn More

CRELD2 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3686

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438314

  • £362.60 / 100µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CRELD2 Polyclonal antibody AF3686 Specifically detects CRELD2 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

CRELD2
LYOPH
Western Blot 0.1 ug/mL
Q9CYA0
cysteine-rich with EGF-like domain protein 2, cysteine-rich with EGF-like domains 2, DKFZp667O055, MGC11256
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse CRELD2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CRELD2 is observed and less than 1% cross-reactivity with recombinant mouse CRELD1 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CRELD2
Mouse myeloma cell line NS0-derived recombinant mouse CRELD2 Ser23-Leu350 Accession # Q9CYA0
Immunogen affinity purified
RUO
Primary
79174
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only