Learn More
CREG Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1697
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487094
  • £382.00 / 100µg
Estimated Shipment
08-03-2018
Add to basket
Description
Description
CREG Polyclonal antibody AF1697 Specifically detects CREG in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
CREG
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
E. coli-derived recombinant mouse CREG, Arg32-Gln220, Accession No. NP_035934
100μg
Primary
433375
Unconjugated
NP_035934
Goat
IgG
RUO
Polyclonal
Murine