missing translation for 'onlineSavingsMsg'
Learn More

Complement Component C1rLP Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3990

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448484

  • £349.86 / 100µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Complement Component C1rLP Polyclonal antibody AF3990 Specifically detects Complement Component C1rLP in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Complement Component C1rLP
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
AAP41417
C1RL, C1RL1, C1RL1complement C1r subcomponent-like protein, C1r-like protein, C1r-like serine protease analog protein, C1r-LPcomplement C1r-like proteinase, C1RLPEC 3.4.21.-, CLSPa, complement component 1, r subcomponent-like, EC 3.4.21, EC 3.4.21.41
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Complement Component C1rLP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human (rh) C1rLP is observed and less than 1% cross-reactivity with rhC1R and rhC1S is observed.
Mouse
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
C1RL
Mouse myeloma cell line NS0-derived recombinant mouse Complement Component C1rLP Ser18-Asp456 Accession # AAP41417
Immunogen affinity purified
RUO
Primary
51279
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only