missing translation for 'onlineSavingsMsg'
Learn More

COCO Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3356

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13428124

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

COCO Polyclonal antibody AF3356 Specifically detects COCO in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

COCO/DAND5
LYOPH
Western Blot 0.1 ug/mL
NP_957679
CER2CRL2, Cerberus 2, Cerberus like-2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DAND5, Dante, DTE, FLJ38607, GREM3, GREM3cerberus 2, Gremlin-3, SP1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse COCO in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human COCO is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
DAND5
E. coli- and mouse myeloma cell line NS0-derived recombinant mouse COCO Arg24-Leu185 Accession # NP_957679
Immunogen affinity purified
RUO
Primary
199699
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only