missing translation for 'onlineSavingsMsg'
Learn More

Chordin-like 2/CHRDL2 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2520

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447614

  • £349.86 / 100µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Chordin-like 2/CHRDL2 Polyclonal antibody AF2520 Specifically detects Chordin-like 2/CHRDL2 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Chordin-like 2/CHRDL2
LYOPH
Western Blot 0.1 ug/mL
Q8VEA6.1
BNF-1, Chordin like 2, chordin-like 2, chordin-like protein 2, CHRDL2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Chordin-like 2/CHRDL2 in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant human Chordin-like 2 is observed and less than 1% cross-reactivity with recombinant mouse (rm) Chordin-like 1 and rmChordin is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CHRDL2
Mouse myeloma cell line NS0-derived recombinant mouse Chordin-like 2/CHRDL2 Gln24-Leu426 Accession # Q8VEA6.1
Immunogen affinity purified
RUO
Primary
25884
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only