missing translation for 'onlineSavingsMsg'
Learn More

Chitinase 3-like 1 Sheep anti-Mouse, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2649

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487694

  • £382.00 / 100µg


Estimated Shipment
05-07-2018
Add to basket

Description

Description

Chitinase 3-like 1 Polyclonal antibody AF2649 Specifically detects Chitinase 3-like 1 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Chitinase 3-like 1
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse Chitinase 3-like 1, Tyr22-Ala381, Accession No. Q61362
100μg
Primary
12654
Unconjugated
Q61362
Sheep
IgG
RUO
Polyclonal
Murine
Videos

For Research Use Only