missing translation for 'onlineSavingsMsg'
Learn More

Cerberus 1 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1986

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487264

  • £377.53 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Cerberus 1 Polyclonal antibody AF1986 Specifically detects Cerberus 1 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Cerberus 1
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse Cerberus 1, Asp41-Pro272, Accession No. O55233
100μg
Primary
12622
Unconjugated
O55233
Goat
IgG
RUO
Polyclonal
Murine
Videos

For Research Use Only