Learn More
CDCP1 Sheep anti-Mouse, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF4515
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488764
  • £377.53 / 100µg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
CDCP1 Polyclonal antibody AF4515 Specifically detects CDCP1 in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
CDCP1
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse CDCP1, Arg25-Ala667, Accession No. Q5U462
100μg
Primary
Murine
Unconjugated
Q5U462
Sheep