missing translation for 'onlineSavingsMsg'
Learn More

CD5L Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2834

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457814

  • £368.97 / 100µg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD5L Polyclonal antibody AF2834 Specifically detects CD5L in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

CD5L
LYOPH
Western Blot 0.1 ug/mL
Q9QWK4
AIM, API6, apoptosis inhibitor 6, CD5 antigen-like, CD5 antigen-like (scavenger receptor cysteine rich family), CD5 molecule-like, CD5L, CT-2, IgM-associated peptide, PRO229, Spalpha, SP-ALPHA
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse CD5L in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CD5L is observed and less than 1% cross-reactivity with recombinant mouse CD5 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CD5L
Mouse myeloma cell line NS0-derived recombinant mouse CD5L Glu22-Val352 Accession # Q9QWK4
Immunogen affinity purified
RUO
Primary
922
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only