missing translation for 'onlineSavingsMsg'
Learn More

CD300b/LMIR5 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2580

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457654

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD300b/LMIR5 Polyclonal antibody AF2580 Specifically detects CD300b/LMIR5 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

CD300b/LMIR5/CD300LB
LYOPH
Western Blot 0.1 ug/mL
Q3U497
CD300 antigen like family member B, CD300 antigen-like family member B, CD300 molecule-like family member b, CD300B, CD300b antigen, CD300LB, CLM7, CLM-7, CLM7IREM3CD300 molecule like family member b, CMRF35A2, CMRF35-A2, CMRF35-like molecule 7, EC 1.13.11.6, EC 6.3.4.3, Gm253, Immune receptor expressed on myeloid cells 3, IREM-3, Leukocyte mono-Ig-like receptor 5, LMIR5, TREM-5, TREM5CD300b, Triggering receptor expressed on myeloid cells 5
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse CD300b/LMIR5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human LMIR5 is observed and less than 5% cross-reactivity with recombinant human (rh) LMIR1, rhLMIR2, recombinant mouse LMIR3, and rhLMIR6 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CD300LB
Mouse myeloma cell line NS0-derived recombinant mouse CD300b/LMIR5 Ile18-Tyr157 Accession # Q3U497
Immunogen affinity purified
RUO
Primary
124599
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only