missing translation for 'onlineSavingsMsg'
Learn More

CD229/SLAMF3 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2555

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467634

  • £332.22 / 100µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD229/SLAMF3 Polyclonal antibody AF2555 Specifically detects CD229/SLAMF3 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

CD229/SLAMF3/Lymphocyte Antigen 9
LYOPH
Western Blot 0.1 ug/mL
AAH95921
CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, Ly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse CD229/SLAMF3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant human CD229 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
LY9
Mouse myeloma cell line NS0-derived recombinant mouse CD229/SLAMF3 Lys48-Phe454 Accession # AAH95921
Immunogen affinity purified
RUO
Primary
4063
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only