missing translation for 'onlineSavingsMsg'
Learn More

Carboxypeptidase A1/CPA1 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2765

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437774

  • £449.92 / 100µg

Estimated Shipment
11-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Carboxypeptidase A1/CPA1 Polyclonal antibody AF2765 Specifically detects Carboxypeptidase A1/CPA1 in Mouse samples. It is validated for Immunoprecipitation, Neutralization, Western Blotting
Specifications

Specifications

Carboxypeptidase A1/CPA1
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 2 ug/mL
Q7TPZ8
carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, CPA1, EC 3.4.17, EC 3.4.17.1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Carboxypeptidase A1/CPA1 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) CPA1 is observed and approximately 10% cross-reactivity with rhCPA2, rhCPB1, and recombinant mouse CPA4 is observed.
Mouse
Western Blot, Immunoprecipitation, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CPA1
Mouse myeloma cell line NS0-derived recombinant mouse Carboxypeptidase A1/CPA1 Asn17-Tyr419 Accession # Q7TPZ8
Immunogen affinity purified
RUO
Primary
1357
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only