missing translation for 'onlineSavingsMsg'
Learn More

Carbonic Anhydrase XII/CA12 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2345

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487494

  • £382.00 / 100µg


Estimated Shipment
05-07-2018
Add to basket

Description

Description

Carbonic Anhydrase XII/CA12 Polyclonal antibody AF2345 Specifically detects Carbonic Anhydrase XII/CA12 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Carbonic Anhydrase XII/CA12
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse Carbonic Anhydrase XII/CA12, Ala25-Ser301, Accession No. Q8CI85
100μg
Primary
76459
Murine
Unconjugated
Q8CI85
Goat
IgG
RUO
Polyclonal
Detects mouse Carbonic Anhydrase XII/CA12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human CA12 is observed and less than 5% cross-reactivity with recombinant human CA1, rhCA2, rhCA3, rhCA8, recombinant mouse (rm) CA4, and rmCA9 is observed.
Videos

For Research Use Only