missing translation for 'onlineSavingsMsg'
Learn More

BOC Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2385

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447524

  • £358.68 / 100µg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

BOC Polyclonal antibody AF2385 Specifically detects BOC in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

BOC
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
AAH78631
Boc homolog (mouse), brother of CDO, Protein BOC
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse BOC in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human BOC is observed.
Mouse
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
BOC
Mouse myeloma cell line NS0-derived recombinant mouse BOC Asp26-Lys508 (Gln ins) Val509-Asp846 Accession # AAH78631
Immunogen affinity purified
RUO
Primary
91653
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only