missing translation for 'onlineSavingsMsg'
Learn More

BCMA/TNFRSF17 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF593

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13469504

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

BCMA/TNFRSF17 Polyclonal antibody AF593 Specifically detects BCMA/TNFRSF17 in Mouse samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications

Specifications

BCMA/TNFRSF17
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
O88472
B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects BCMA/TNFRSF17 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human BCMA is observed.
Mouse
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TNFRSF17
Mouse myeloma cell line NS0-derived recombinant mouse BCMA/TNFRSF17 Met1-Thr49 Accession # O88472
Immunogen affinity purified
RUO
Primary
608
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only