missing translation for 'onlineSavingsMsg'
Learn More

Axl Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF854

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13450275

  • £433.65 / 100µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Axl Polyclonal antibody AF854 Specifically detects Axl in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Axl
LYOPH
Western Blot 0.1 ug/mL
Q80YQ3
ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Axl in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human Axl is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
AXL
Mouse myeloma cell line NS0-derived recombinant mouse Axl His20-Pro443 Accession # Q80YQ3
Antigen Affinity-purified
RUO
Primary
558
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only