Learn More
ASCL1/Mash1 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2567
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13457644
  • £368.97 / 100µg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
ASCL1/Mash1 Polyclonal antibody AF2567 Specifically detects ASCL1/Mash1 in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
ASCL1/Mash1
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Q53WV9
achaete-scute complex (Drosophila) homolog-like 1, achaete-scute complex homolog 1 (Drosophila), achaete-scute complex-like 1 (Drosophila), achaete-scute homolog 1, ASCL1, ASH1, ASH-1, ASH1MASH1, BHLHA46, bHLHa46achaete-scute complex-like 1, Class A basic helix-loop-helix protein 46, HASH1, HASH1achaete scute protein, Mash1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse ASCL1/Mash1 in direct ELISAs and Western blots.
Mouse
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ASCL1
E. coli-derived recombinant mouse ASCL1/Mash1 His74-Asp173 (Gln160Glu) Accession # Q53WV9
Antigen Affinity-purified
RUO
Primary
429
Reconstitute at 0.2 mg/mL in sterile PBS.