missing translation for 'onlineSavingsMsg'
Learn More

ASAH2/N-acylsphingosine Amidohydrolase-2 Sheep anti-Mouse, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3558

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488234

  • £347.17 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

ASAH2/N-acylsphingosine Amidohydrolase-2 Polyclonal antibody AF3558 Specifically detects ASAH2/N-acylsphingosine Amidohydrolase-2 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

ASAH2/N-acylsphingosine Amidohydrolase-2
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q9JHE3
ASAH2 N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2, BCDase, HNAC1, LCDase, Nacylsphingosine Amidohydrolase2, NCDase, N-CDase
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse ASAH2/N-acylsphingosine Amidohydrolase-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human ASAH2 is observed.
Mouse
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ASAH2
Mouse myeloma cell line NS0-derived recombinant mouse ASAH2/N-acylsphingosine Amidohydrolase-2, Thr34-Thr756, Accession # Q9JHE3
Immunogen affinity purified
RUO
Primary
56624
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only