missing translation for 'onlineSavingsMsg'
Learn More

Arylsulfatase G/ARSG Sheep anti-Mouse, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4600

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448814

  • £349.86 / 100µg

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Arylsulfatase G/ARSG Polyclonal antibody AF4600 Specifically detects Arylsulfatase G/ARSG in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Arylsulfatase G/ARSG
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q3TYD4
ARSG, arylsulfatase G, ASG, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.4, KIAA1001ASG
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Arylsulfatase G/ARSG in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant mouse ARSA, recombinant human (rh) ARSB and rhARSJ is observed.
Mouse
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ARSG
Chinese hamster ovary cell line CHO-derived recombinant mouse Arylsulfatase G/ARSG Gly17-Val525 Accession # Q3TYD4
Immunogen affinity purified
RUO
Primary
22901
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only