missing translation for 'onlineSavingsMsg'
Learn More

APLP-2 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3685

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13428314

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

APLP-2 Polyclonal antibody AF3685 Specifically detects APLP-2 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

APLP-2
LYOPH
Western Blot 0.1 ug/mL
AAH52396
amyloid beta (A4) precursor-like protein 2, amyloid precursor protein homolog HSD-2, Amyloid protein homolog, amyloid-like protein 2, APLP2, APLP-2, APPH, APPHCDEI box-binding protein, APPL2, CDEBP, WALPLP2, YWK-II
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse APLP-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) APLP-2 is observed and less than 1% cross-reactivity with rhAPLP-1 and recombinant mouse APLP-1 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
APLP2
Mouse myeloma cell line NS0-derived recombinant mouse APLP-2 Leu37-Ser624 Accession # AAH52396
Immunogen affinity purified
RUO
Primary
334
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only