missing translation for 'onlineSavingsMsg'
Learn More

APLP-1 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3179

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458024

  • £298.33 / 100µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

APLP-1 Polyclonal antibody AF3179 specifically detects APLP-1 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

APLP-1
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q03157
amyloid beta (A4) precursor-like protein 1, amyloid precursor-like protein 1, amyloid-like protein 1, APLP1, APLPAPLP-1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse APLP-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant human APLP-1 is observed and less than 1% cross-reactivity with recombinant mouse APLP-2 is observed.
Mouse
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
DPP4
Mouse myeloma cell line NS0-derived recombinant mouse APLP-1 Gly37-Arg582 Accession # Q03157
Immunogen affinity purified
RUO
Primary
333
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only