missing translation for 'onlineSavingsMsg'
Learn More

Aminopeptidase A/ENPEP Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2809

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13427804

  • £332.22 / 100µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Aminopeptidase A/ENPEP Polyclonal antibody AF2809 Specifically detects Aminopeptidase A/ENPEP in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Aminopeptidase A/ENPEP
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
P16406
Aminopeptidase A, APA, AP-A, CD249, CD249 antigen, Differentiation antigen gp160, EAP, EC 3.4.11, EC 3.4.11.7, ENPEP, glutamyl aminopeptidase, glutamyl aminopeptidase (aminopeptidase A), gp160
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Aminopeptidase A/ENPEP in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human ENPEP is observed.
Mouse
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ENPEP
Mouse myeloma cell line NS0-derived recombinant mouse Aminopeptidase A/ENPEP Arg41-Pro945 Accession # P16406
Immunogen affinity purified
RUO
Primary
2028
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only