missing translation for 'onlineSavingsMsg'
Learn More

AMIGO3 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2375

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457514

  • £332.22 / 100µg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

AMIGO3 Polyclonal antibody AF2375 Specifically detects AMIGO3 in Mouse samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

AMIGO3
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Q8C2S7
adhesion molecule with Ig-like domain 3, ALI3, Alivin-3, AMIGO-3, amphoterin-induced gene and ORF 3, amphoterin-induced protein 3, KIAA1851, MGC120552
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse AMIGO3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant mouse AMIGO2 is observed and less than 2% cross-reactivity with recombinant human AMIGO1 is observed.
Mouse
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
AMIGO3
Mouse myeloma cell line NS0-derived recombinant mouse AMIGO3 Thr20-Pro378 Accession # Q8C2S7
Antigen Affinity-purified
RUO
Primary
386724
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only