missing translation for 'onlineSavingsMsg'
Learn More

AMIGO2 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2374

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447514

  • £332.22 / 100µg

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

AMIGO2 Polyclonal antibody AF2374 Specifically detects AMIGO2 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

AMIGO2
LYOPH
Western Blot 0.1 ug/mL
Q80ZD9
adhesion molecule with Ig-like domain 2, ALI1amphoterin induced gene 2, alivin 1, Alivin-1, AMIGO-2, DEGAamphoterin-induced protein 2, differentially expressed in gastric adenocarcinoma, Differentially expressed in gastric adenocarcinomas, transmembrane protein AMIGO2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse AMIGO2 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) AMIGO2 is observed and less than 1% cross-reactivity with rhAMIGO1 and recombinant mouse AMIGO3 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
AMIGO2
Mouse myeloma cell line NS0-derived recombinant mouse AMIGO2 Gy38-Ala391 Accession # Q80ZD9
Immunogen affinity purified
RUO
Primary
347902
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only