Learn More
ADAM15 Ectodomain Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF945
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13460345
  • £314.17 / 100µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
ADAM15 Ectodomain Polyclonal antibody AF945 Specifically detects ADAM15 Ectodomain in Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Specifications
ADAM15
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
O88839
ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC15, MDC-15, Metargidin
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse ADAM15 Ectodomain in direct ELISAs and Western blots. In Western blots (under reducing and non-reducing conditions), no cross-reactivity with the ectodomains of recombinant mouse (rm) ADAM9, rmADAM10, recombinant human (rh) ADAM8, rhADAM15, and rhADAM17 is observed.
Mouse
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ADAM15
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ADAM15 Arg18-Ser694 Accession # O88839
Antigen Affinity-purified
RUO
Primary
8751
Reconstitute at 0.2 mg/mL in sterile PBS.