missing translation for 'onlineSavingsMsg'
Learn More

2B4/CD244/SLAMF4 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13466694

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

2B4/CD244/SLAMF4 Polyclonal antibody AF1050 Specifically detects 2B4/CD244/SLAMF4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blotting
Specifications

Specifications

2B4/CD244/SLAMF4
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready
Q07763
CD244, CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAIL, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4, SLAMF4NK cell activation inducing ligand NAIL
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse 2B4/CD244/SLAMF4 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) 2B4 is observed and less than 1% cross-reactivity with rhSLAM is observed.
Mouse
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CD244
Mouse myeloma cell line NS0-derived recombinant mouse 2B4/CD244/SLAMF4 His17-Asn221 Accession # Q07763
Immunogen affinity purified
RUO
Primary
51744
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only