Learn More
Pro MMP-9 Rat anti-Mouse, Clone: 116134, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB9092
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13461976
  • £390.72 / 500µg
Estimated Shipment
14-09-2020
Description
Description
Pro MMP-9 Monoclonal antibody MAB9092 Specifically detects Pro MMP-9 in Mouse samples. It is validated for ELISA Capture
Specifications
Specifications
MMP-9
116134
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P41245.1
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Pro-MMP-9 in ELISAs. This antibody does not recognize the mature form of Pro-MMP-9.
Mouse
ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
MMP9
Mouse myeloma cell line NS0-derived recombinant mouse MMP-9 Ala20-Pro730 Accession # P41245.1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4318
Reconstitute at 0.5 mg/mL in sterile PBS.