missing translation for 'onlineSavingsMsg'
Learn More

MMP-25/MT6-MMP Mouse anti-Human, Clone: 141808, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1142

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488385

  • £353.77 / 500µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

MMP-25/MT6-MMP Monoclonal antibody MAB1142 Specifically detects MMP-25/MT6-MMP in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

MMP-25/MT6-MMP
141808
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
MMP25
Mouse myeloma cell line NS0-derived recombinant human MMP-25/MT6-MMP, Pro314-Cys508, Accession # Q9NPA2
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
64386
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, CyTOF
LYOPH
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q9NPA2
leukolysin, matrix metallopeptidase 25, matrix metallopeptidase-like 1, matrix metalloproteinase 20, matrix metalloproteinase 25, matrix metalloproteinase-25, matrix metalloproteinase-like 1, membrane-type 6 matrix metalloproteinase, membrane-type matrix metalloproteinase 6, membrane-type-6 matrix metalloproteinase, MMP20, MMP20A, MMP25, MMP-25, MMPL1, MT6MMP, MT6-MMP, MT-MMP 6, MTMMP6, MT-MMP6
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human MMP-25/MT6-MMP in Western blots.
Human
Videos

For Research Use Only