Learn More
Pro MMP-13 Mouse anti-Human, Clone: 87518, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB913
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13451986
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
Pro MMP-13 Monoclonal antibody MAB913 Specifically detects Pro MMP-13 in Human samples. It is validated for Affinity Purification, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Specifications
MMP-13
87518
Western Blot 1 ug/mL, ELISA, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunoaffinity Purification
P45452
CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP13, MMP-13
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Pro MMP-13 in direct ELISAs and Western blots. Does not detect active recombinant human MMP-13. In Western blots, no cross-reactivity with recombinant human MMP-1, -2, -3, -7, -8, -9, -10, or -12 is observed.
Human
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Affinity Purification
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
MMP13
Chinese hamster ovary cell line CHO-derived recombinant human MMP-13 Leu20-Cys471 Accession # P45452
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4322
Reconstitute at 0.5 mg/mL in sterile PBS.