Learn More
MMP-12 Catalytic Domain Mouse anti-Human, Clone: 82902, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB919
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13441996
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
MMP-12 Catalytic Domain Monoclonal antibody MAB919 Specifically detects MMP-12 Catalytic Domain in Human samples. It is validated for Affinity Purification, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Specifications
MMP-12
82902
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunoaffinity Purification
Purified
MMP12
Chinese hamster ovary cell line CHO-derived recombinant human MMP-12
Protein A or G purified from ascites
RUO
Primary
4321
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunohistochemistry, Immunoprecipitation, Affinity Purification
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
EC 3.4.24, hME, HMEEC 3.4.24.65, Macrophage elastase, macrophage metalloelastase, matrix metallopeptidase 12 (macrophage elastase), matrix metalloproteinase 12 (macrophage elastase), Matrix metalloproteinase-12, ME, MGC138506, MME, MMP12, MMP-12
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human MMP-12 Catalytic Domain. Does not recognize the C-terminal hemopexin-like domain of human MMP-12. Does not cross-react with recombinant human MMP-1, -2, -3, -7, -8, -9, -10, or -13.