Learn More
MMP-11 Mouse anti-Human, Clone: 135421, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3657
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13430196
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
MMP-11 Monoclonal antibody MAB3657 Specifically detects MMP-11 in Human samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
Specifications
MMP-11
135421
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
MMP11
E. coli-derived recombinant human MMP-11 Met1-Leu488 Accession # P24347
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4320
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, CyTOF
LYOPH
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
P24347
EC 3.4.24, EC 3.4.24.-, EC 3.4.24.24, matrix metallopeptidase 11 (stromelysin 3), matrix metalloproteinase 11 (stromelysin 3), Matrix metalloproteinase-11, MMP11, MMP-11, SL-3, ST3, STMY3stromelysin 3, stromelysin III, stromelysin-3
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human MMP-11 in direct ELISAs.